Alfa Aesar™ 4-Trimethylsilyl-3-butyn-1-ol, 98%

Manufacturer: Alfa Aesar™ H5337606

Quantity 5g

 View more versions of this product

Catalog No. AAH5337606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2117-12-6
    Molecular Formula C7H14OSi
    Linear Formula HOCH2CH2C≡CSi(CH3)3
    Formula Weight 142.27
    MDL Number MFCD00190217
    UN Number UN1993
    Chemical Name or Material 4-Trimethylsilyl-3-butyn-1-ol

    Specifications

    Formula Weight 142.27
    Molecular Formula C7H14OSi
    Linear Formula HOCH2CH2C≡CSi(CH3)3
    MDL Number MFCD00190217
    UN Number UN1993
    Boiling Point 97° to 98°C (24mmHg)
    Quantity 5g
    Flash Point 60°C (140°F)
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 2117-12-6
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Chemical Name or Material 4-Trimethylsilyl-3-butyn-1-ol
    Assay 98%

    Description

    Applications
    4-Trimethylsilyl-3-butyn-1-ol is used as an organic chemical synthesis intermediate.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit