Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Trimethylsilyl-3-butyn-1-ol, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2117-12-6
C7H14OSi
142.27
MFCD00190217
VSTMVAFCUIGHQI-UHFFFAOYSA-N
4-trimethylsilyl-3-butyn-1-ol,4-trimethylsilyl but-3-yn-1-ol,3-butyn-1-ol, 4-trimethylsilyl,4-trimethylsilyl-3-butyne-1-ol,1-trimethylsilyl-1-butyn-4-ol,3-butyn-1-ol,4-trimethylsilyl
2760827
4-trimethylsilylbut-3-yn-1-ol
C[Si](C)(C)C#CCCO
2117-12-6
C7H14OSi
142.27
MFCD00190217
VSTMVAFCUIGHQI-UHFFFAOYSA-N
4-trimethylsilyl-3-butyn-1-ol,4-trimethylsilyl but-3-yn-1-ol,3-butyn-1-ol, 4-trimethylsilyl,4-trimethylsilyl-3-butyne-1-ol,1-trimethylsilyl-1-butyn-4-ol,3-butyn-1-ol,4-trimethylsilyl
2760827
4-trimethylsilylbut-3-yn-1-ol
C[Si](C)(C)C#CCCO
Infrared Spectrum Authentic
Refractive Index 1.4540 to 1.4560 (20°C, 589nm)
Density 0.8540g/mL
Packaging Glass bottle
Specific Gravity 0.854
Quantity 5g
Boiling Point 97.0°C to 98.0°C (24.0mmHg)
Color Black-Brown to Gray
Flash Point 60°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula (CH3)3SiC≡C(CH2)2OH
Formula Weight 142.27
Physical Form Solution
Percent Purity 98%
Chemical Name or Material 4-Trimethylsilyl-3-butyn-1-ol
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning