Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Trimethylsilyl)morpholine, 96%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
13368-42-8
C7H17NOSi
159.304
MFCD00010098
JJOWIQMPCCUIGA-UHFFFAOYSA-N
4-trimethylsilyl morpholine,n-trimethylsilyl morpholine,n-trimethylsilylmorpholine,trimethyl morpholin-4-yl silane,morpholine, 4-trimethylsilyl,trimethylsilyl morpholin,morpholinotrimethylsilane,trimethylmorpholinosilane,acmc-1btin,timtec-bb sbb009054
559763
trimethyl(morpholin-4-yl)silane
C[Si](C)(C)N1CCOCC1
13368-42-8
C7H17NOSi
159.304
MFCD00010098
JJOWIQMPCCUIGA-UHFFFAOYSA-N
4-trimethylsilyl morpholine,n-trimethylsilyl morpholine,n-trimethylsilylmorpholine,trimethyl morpholin-4-yl silane,morpholine, 4-trimethylsilyl,trimethylsilyl morpholin,morpholinotrimethylsilane,trimethylmorpholinosilane,acmc-1btin,timtec-bb sbb009054
559763
trimethyl(morpholin-4-yl)silane
C[Si](C)(C)N1CCOCC1
Quantity 25g
Refractive Index 1.4410 to 1.4430 (20°C, 589nm)
Infrared Spectrum Authentic
Color Colorless to Yellow
Density 0.897g/mL
Packaging Glass bottle
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Formula Weight 159.3
Physical Form Liquid
Percent Purity ≥95%
Chemical Name or Material 4-(Trimethylsilyl)morpholine
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger