Alfa Aesar™ 4-(Trimethylsilyl)morpholine, 97%

Manufacturer: Alfa Aesar™ H5522903

Quantity 1g

 View more versions of this product

Catalog No. AAH5522903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13368-42-8
    Molecular Formula C7H17NOSi
    Formula Weight 159.3
    MDL Number MFCD00010098
    UN Number UN2924
    Beilstein 1071480
    Chemical Name or Material 4-(Trimethylsilyl)morpholine

    Specifications

    Formula Weight 159.3
    Molecular Formula C7H17NOSi
    MDL Number MFCD00010098
    CAS 13368-42-8
    UN Number UN2924
    Beilstein 1071480
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318-H226
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    Flash Point 29°C (84°F)
    Boiling Point 160°C
    Quantity 1g
    Chemical Name or Material 4-(Trimethylsilyl)morpholine
    Assay 97%

    GHS H Statement
    H314-H318-H226
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit