Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Trimethylsilyloxy-3-penten-2-one 96.0+%, TCI America™

Click to view available options
Quantity:
25mL
13257-81-3
C8H16O2Si
172.299
MFCD00009853
FBADCSUQBLLAHW-SOFGYWHQSA-N
6059369
(E)-4-trimethylsilyloxypent-3-en-2-one
CC(=CC(=O)C)O[Si](C)(C)C
13257-81-3
C8H16O2Si
172.299
MFCD00009853
FBADCSUQBLLAHW-SOFGYWHQSA-N
6059369
(E)-4-trimethylsilyloxypent-3-en-2-one
CC(=CC(=O)C)O[Si](C)(C)C
Color Yellow
Boiling Point 58°C
Quantity 25mL
UN Number 1993
Formula Weight 172.30
Physical Form Clear Liquid at 20°C
Percent Purity ≥96.0% (GC)
Chemical Name or Material 4-Trimethylsilyloxy-3-penten-2-one