Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Tritylphenol, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
978-86-9
C25H20O
336.434
MFCD00002364
NIPKXTKKYSKEON-UHFFFAOYSA-N
4-triphenylmethylphenol,4-triphenylmethyl phenol,p-tritylphenol,p-triphenylmethylphenol,phenol, 4-triphenylmethyl,p-cresol, .alpha.,.alpha.,.alpha.-triphenyl,4-tritylphenol,p-triphenylmethyl phenol,4-tri phenyl methyl phenol,phenol,4-triphenylmethyl
70422
4-tritylphenol
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=C(C=C4)O
978-86-9
C25H20O
336.434
MFCD00002364
NIPKXTKKYSKEON-UHFFFAOYSA-N
4-triphenylmethylphenol,4-triphenylmethyl phenol,p-tritylphenol,p-triphenylmethylphenol,phenol, 4-triphenylmethyl,p-cresol, .alpha.,.alpha.,.alpha.-triphenyl,4-tritylphenol,p-triphenylmethyl phenol,4-tri phenyl methyl phenol,phenol,4-triphenylmethyl
70422
4-tritylphenol
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=C(C=C4)O
Quantity 5g
Color Yellow
Melting Point 283°C to 289°C
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 06, 731
Formula Weight 336.43
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 4-Tritylphenol, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 213-557-1