Alfa Aesar™ 4-Vinylbenzeneboronic acid, 97%

Manufacturer: Alfa Aesar™ B2370906

Quantity 5g

 View more versions of this product

Catalog No. AAB2370906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2156-04-9
    Molecular Formula C8H9BO2
    Formula Weight 147.97
    MDL Number MFCD00239441
    Beilstein 2829387
    Synonym Styrene-4-boronic acid; 4-Vinylphenylboronic acid
    Chemical Name or Material 4-Vinylbenzeneboronic acid

    Specifications

    Formula Weight 147.97
    Molecular Formula C8H9BO2
    MDL Number MFCD00239441
    Synonym Styrene-4-boronic acid; 4-Vinylphenylboronic acid
    Melting Point ≈190°C
    Quantity 5g
    Recommended Storage 2° to 8°C
    CAS 2156-04-9
    Beilstein 2829387
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 4-Vinylbenzeneboronic acid
    Assay 97%

    Description

    Applications
    4-Vinylbenzeneboronic acid is widely used in Suzuki reaction. It is also used in the preparation of boronic acid containing polymers, which is used in potential biomedical applications.

    Solubility
    Soluble in ethanol, dimethylsulfoxide, tetrahydrofuran and diethyl ether. Slightly soluble in water.

    Notes
    Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2° to 8°C

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit