Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Vinylbenzeneboronic acid, 97%

Click to view available options
Quantity:
1g
5g
25g
2156-04-9
C8H9BO2
147.97
MFCD00239441
QWMJEUJXWVZSAG-UHFFFAOYSA-N
4-vinylphenylboronic acid,4-vinylbenzeneboronic acid,4-vinylphenyl boronic acid,4-ethenylphenyl boronic acid,4-vinylboronic acid,p-styrylboronic acid,p-vinylphenylboronic acid,styrene-4-boronic acid,boronic acid, 4-ethenylphenyl,4-styreneboronic acid
2734393
(4-ethenylphenyl)boronic acid
B(C1=CC=C(C=C1)C=C)(O)O
2156-04-9
C8H9BO2
147.97
MFCD00239441
QWMJEUJXWVZSAG-UHFFFAOYSA-N
4-vinylphenylboronic acid,4-vinylbenzeneboronic acid,4-vinylphenyl boronic acid,4-ethenylphenyl boronic acid,4-vinylboronic acid,p-styrylboronic acid,p-vinylphenylboronic acid,styrene-4-boronic acid,boronic acid, 4-ethenylphenyl,4-styreneboronic acid
2734393
(4-ethenylphenyl)boronic acid
B(C1=CC=C(C=C1)C=C)(O)O
Applications
4-Vinylbenzeneboronic acid is widely used in Suzuki reaction. It is also used in the preparation of boronic acid containing polymers, which is used in potential biomedical applications.

Solubility
Soluble in ethanol, dimethylsulfoxide, tetrahydrofuran and diethyl ether. Slightly soluble in water.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Quantity 1g
Melting Point ~190°C
Beilstein 2829387
Formula Weight 147.97
Percent Purity 97%
Chemical Name or Material 4-Vinylbenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C