Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Vinylbenzeneboronic acid, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
2156-04-9
C8H9BO2
147.97
QWMJEUJXWVZSAG-UHFFFAOYSA-N
4-vinylphenylboronic acid,4-vinylbenzeneboronic acid,4-vinylphenyl boronic acid,4-ethenylphenyl boronic acid,4-vinylboronic acid,p-styrylboronic acid,p-vinylphenylboronic acid,styrene-4-boronic acid,boronic acid, 4-ethenylphenyl,4-styreneboronic acid
2734393
(4-ethenylphenyl)boronic acid
B(C1=CC=C(C=C1)C=C)(O)O
2156-04-9
C8H9BO2
147.97
QWMJEUJXWVZSAG-UHFFFAOYSA-N
4-vinylphenylboronic acid,4-vinylbenzeneboronic acid,4-vinylphenyl boronic acid,4-ethenylphenyl boronic acid,4-vinylboronic acid,p-styrylboronic acid,p-vinylphenylboronic acid,styrene-4-boronic acid,boronic acid, 4-ethenylphenyl,4-styreneboronic acid
2734393
(4-ethenylphenyl)boronic acid
B(C1=CC=C(C=C1)C=C)(O)O
Melting Point 190°C
Packaging Glass bottle
Color White to Beige
Quantity 1g
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 16, IV, 1676
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,dmso,dmf,thf,diethyl ether
Formula Weight 147.97
Physical Form Crystalline Powder or Crystals
Percent Purity 98%
Chemical Name or Material 4-Vinylbenzeneboronic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning