Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Vinylpyridine, Stabilized 95%, ACROS Organics™

Click to view available options
Quantity:
100mL
250mL
1L
Packaging:
Glass bottle
100-43-6,123-31-9
C7H7N
105.14
MFCD00006447
KFDVPJUYSDEJTH-UHFFFAOYSA-N
4-vinylpyridine,pyridine, 4-ethenyl,pyridine, 4-vinyl,gamma-vinylpyridine,unii-i56g67xm8d,ccris 5240,4vp,4-pyridylethylene,4-vinyl-pyridine,4-vinyl pyridine
7502
4-ethenylpyridine
C=CC1=CC=NC=C1
100-43-6,123-31-9
C7H7N
105.14
MFCD00006447
KFDVPJUYSDEJTH-UHFFFAOYSA-N
4-vinylpyridine,pyridine, 4-ethenyl,pyridine, 4-vinyl,gamma-vinylpyridine,unii-i56g67xm8d,ccris 5240,4vp,4-pyridylethylene,4-vinyl-pyridine,4-vinyl pyridine
7502
4-ethenylpyridine
C=CC1=CC=NC=C1
Boiling Point 171°C
Melting Point -20°C
Viscosity <3 cps (20°C)
Color Undesignated
Infrared Spectrum Authentic
Density 0.9890g/mL
Quantity 1L
Flash Point 48°C
Specific Gravity 0.989
Stabilizer 80 to 150ppm hydroquinone
Refractive Index 1.5500 to 1.5520
Packaging Glass bottle
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula (C5H4N)CH=Ch2
Beilstein 20, II, 170
Solubility Information Solubility in water: 29g/L (20°C). Other solubilities: soluble in ethanol,chloroform,in diluted acids,slightly soluble in diethyl ether
Formula Weight 105.14
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 4-Vinylpyridine, 95%
Show More Show Less

GHS H Statement
Flammable liquid and vapor. Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes severe skin burns and eye damage. May cause an allergic skin reaction. Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

Danger

EINECSNumber : 202-852-0

RTECSNumber : UU1045000

TSCA : TSCA

Recommended Storage : Product may darken during storage