Learn More
Aviva Systems Biology Corp SEPT2 Antibody - N-terminal region (ARP56660_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP56660_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SEPT2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SEPT2. Alternate names: DIFF6, KIAA0158, NEDD5, Pnutl3, hNedd5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61943