Learn More
Aviva Systems Biology Corp SEPT12 Antibody - middle region (ARP52697_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP52697_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SEPT12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SEPT12. Alternate names: FLJ25410ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-60807