Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Wheaton™ Safety-Coated Reagent Bottles
Click to view available options
Capacity (Metric):
250 mL
Description
Features convenient writing patch.
- Clear borosilicate glass
- Attached 33-430 screw cap
- Polypropylene cap with PTFE liner
- Autoclavable
Specifications
Closure Material | Polypropylene |
Closure Size | 33-430 mm |
Closure Type | Screw Cap |
Closure Liner | Polyethylene Foam |
Diameter (Metric) | 69 mm |
Height (Metric) | 151 mm |
Capacity | 8.4 oz., 250mL |
Capacity (Metric) | 250 mL |
Capacity (English) | 8.4 oz. |
Color | Clear |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction