Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ Blue 1H Half-Mask Respirators
Half-mask respirators with P100 filters are designed specifically for healthcare workers.
Supplier: Honeywell™ B210080
Includes: One pair of P100 particulate filters and filter splash guards.
Description
Reusable respirator to protect healthcare workers from the hazards and challenges faced each day in infectious disease environments
- Includes a speech diaphragm for better worker-to-patient speech clarity for improved communication
- Splash guards for the P100 filters help prevent blood borne pathogens from contaminating the exterior of the filters
- Splash guard over the exhalation port directs healthcare worker's exhaled breath away from patients
- Silicone facepiece is soft and flexible to provide workers with a comfortable fit for hours of use
- Dual sealing flange provides excellent facepiece seal
- Cradle suspension provides excellent support for a secure fit
- NIOSH approved (with P100 particulate filters only)
Specifications
P100 Cartridge | |
Lilac | |
Half-Mask Respirator | |
Healthcare professionals in environments ranging from the ER, OR, and IC to clinics and other medical facilities | |
No | |
Cradle |
Threaded | |
Silicone | |
Small | |
P100 filters and splash guards | |
B210080 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction