Learn More
Brady™ Oil Only Socks
Dependable, flexible tubes that contain and absorb liquids
$213.00 - $323.00
Specifications
Material | Polypropylene |
---|---|
Type | Oil Only Socks |
For Use With (Application) | For use against oil and petroleum based spills |
Catalog Number | Mfr. No. | Packaging Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Packaging Quantity | Price | Quantity & Availability | |||||
19-000-182
|
Brady™
OIL124 |
4/Cs. |
Case of 4 for $213.00
|
|
|||||
19-000-185
|
Brady™
OIL806 |
6/Cs. |
Case of 6 for $217.00
|
|
|||||
19-000-183
|
Brady™
OIL412 |
12/Cs. |
Case of 12 for $221.00
|
|
|||||
19-000-184
|
Brady™
OIL430 |
30/Cs. |
Case of 30 for $323.00
|
|
|||||
Description
- Made of oil-only polypropylene SOC
- Molds around corners and conforms to uneven surfaces
- Does not absorb water or water-based solutions
- Wringable and can be incinerated
- Used as a working tool for all major industries
- Available in variety of types and sizes
Specifications
Polypropylene | |
Oil Only Socks | |
For use against oil and petroleum based spills |