Learn More
Scott Safety™ ACSi SCBA
Offers the Advanced Carrying System (ACS), which features a padded Thermoflex backplate molded to fit the spine and back muscles for increased comfort and flexibility
Manufacturer:  Scott Safety™ ACS3A2013000101
 View more versions of this product
Catalog No. 19-122-028
Add to cart
Description
Description
  • Lightweight, flexible Thermoflex backplate for greater comfort and flexibility
  • EZ-Flo regulator reduces inhalation and exhalation resistance for reduced user fatigue
  • Pneumatics are easily removed with a Scott “quarter-turn”—no tools required—for ease of maintenance and turn-around
  • Adjustable, padded shoulder straps and waistband with quick-release buckle for comfort and ease of donning and doffing
  • Shoulder mounted photoluminescent gauge with whistle end-of-service alarm for ease of use
  • Cylinder: 2216 psi Aluminum, 30 min.
Specifications
Specifications
2216 psig
None
Cylinder: 2216 psi Aluminum, 30 min.; Manifold: Standard
ACSI Standard
None
EZ-Flo Standard