Learn More
Scott Safety™ AIR-PAK 75i SCBAs
Designed with durability, dependability for insudtrial applications
Supplier: Scott Safety™ API452002053111
Description
- Streamlined backframe provides comfortable fit with lower profile
- Featues one-piece backframe constructed of lightweight, time-tested, durable aluminum alloy
- Parachute-Style, oversized harness fasteners designed with gloved hands
- Enhanced latching system with durable para-aramid fabric band
- Includes efficient breathing circuit
- Features Top Down Convertibility concept for c increase in versatility, compatibility with Scott product line
- E-Z Flo regulator with Vibralert end-of-service indicator
- Has two dual-redundant pressure reducer interconnected through automatic transfer valve
- Quick-don mounting system in E-Z Flo regulator
- Air-saver switch angled for easy operation with gloved hand
- Fully padded shoulder straps and waist belt
- Integrated donning/carrying handles
- Dual mechanical speaking diaphragms
- Remote pressure gauge
- Cam-Over center cylinder adjustment band
- Drag rescue loop
Specifications
SCBA | |
AV-3000 Sureseal Dupont™ Kevlar™ aramid fibers (M) | |
Airline option:Hansen HK socket; |
2216 psig | |
Industrial applications | |
E-Flo regulator w/Vibralert Quick Connect |