Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversified Spaces™ Stainless-Steel Tables
SCI_ED

Diversified Stainless-Steel Table features a top with integrally welded C channels to prevent sagging.

Supplier:  Diversified Spaces™ XS3630

 View more versions of this product

Catalog No. S14188


Add to Cart

Description

Description

  • Stainless-steel top with two sound deadened integrally welded 12.7cm (5 in.) channels to reinforce and maintain level working surface (prevents sags)
  • Material: T-430 18-gauge stainless-steel
  • Galvanized legs (4.1cm dia.) with adjustable high impact corrosion resistant feet
  • Adjustable under-shelf with two pins per corner provides strength and stability
Specifications

Specifications

NSF
76.2 cm
89 cm
36 in.
Table
30 in.
35 in.
Stainless Steel
91.4 cm
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit