Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diversified Spaces™ Stainless-Steel Tables

Diversified Stainless-Steel Table features a top with integrally welded C channels to prevent sagging.
Supplier: Diversified Spaces™ XS4830
Description
- Stainless-steel top with two sound deadened integrally welded 12.7cm (5 in.) channels to reinforce and maintain level working surface (prevents sags)
- Material: T-430 18-gauge stainless-steel
- Galvanized legs (4.1cm dia.) with adjustable high impact corrosion resistant feet
- Adjustable under-shelf with two pins per corner provides strength and stability
Specifications
NSF | |
76.2 cm | |
89 cm | |
48 in. | |
Table |
30 in. | |
35 in. | |
Stainless Steel | |
122 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title