Learn More
Abbott iCup™ Dx Pro Urine Drug Screening Device
Easy to use, non-tilt auto activated cup for screening a wide range of drugs
Manufacturer: Abbott IDXP111701
This item is not returnable.
View return policy
Description
- Self-contained cup is ideal for sending presumptive positive specimens to lab for confirmation
- Low-level opiate cutoffs (300ng/mL)
- Available with specimen validity test (SVT) and temperature strip help to monitor specimen tampering (SVT includes SG, OX, pH)
- Simple to use device gives results in five minutes
- CLIA waived
Specifications
AMP, BAR, BZO, COC, mAMP, MDMA, MTD, MOP(OPI300), OXY, THC, TCA (OX/SG/PH) |