Learn More
Hamilton™ Microlab™ 1700 Series Gastight™ Syringes
For dispensing both liquids and gases
$80.30 - $84.70
Catalog Number | Mfr. No. | Volume (Metric) | Model | Needle Gauge | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Model | Needle Gauge | Price | Quantity & Availability | |||
14-824-677
|
Hamilton™
80262 |
25 μL | 1702 | 22s G |
Each for $84.70
|
|
14-824-678
|
Hamilton™
81062 |
100 μL | 1710 | 22s G |
Each for $84.70
|
|
14-824-676
|
Hamilton™
80062 |
10 μL | 1701 | 26s G |
Each for $80.30
|
|
Description
- Equipped with precision-machined polymer plunger tip, often PTFE, which creates leak-free seal
- Tight fit feature ensures tip wipes interior of syringe barrel free of sample
- Reduces the chance deposit will occur in samples with dissolved or suspended molecules and result in damaged plunger
- ChemSeal termination