Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BrandTech™ BRAND™ Reservoir Bottles for Automatic Burette

Spare 2000mL bottles for automatic burette manufactured of boro.3.3 glass.

Manufacturer:  BrandTech™ BRAND™ 23320

 View more versions of this product

Catalog No. 13-688-991


Add to cart

Description

Description

  • Manufactured of boro 3.3 glass with a PP screw-cap and PE-LD pouring ring
  • Maximum operating temperature is 80°C (cleaning temperatures should not exceed 60°C)
  • Requires use of bottletop adapter - 33mm/STJ 29/32
Specifications

Specifications

Amber
Automatic Burette
Reservoir Bottle
2000 mL
Borosilicate Glass
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit