Learn More
Brady™ SPC™ MAXX Enhanced Heavy Weight Oil Rolls
For use with oil and petroleum based fluids
$315.00 - $341.00
Specifications
Type MAXX Enhanced Heavy Weight Roll
For Use With (Application) Overspray leaks and drips, workstations, drums - liquid storage, outfalls and discharge Ponds
View More Specs
Products 2
Catalog Number Mfr. No. Dimensions (L x W) Capacity (English) Packaging Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Capacity (English) Packaging Quantity Price Quantity & Availability  
17-111-175
View Documents
Brady™
SPC150
43.8 m x 96.5 cm (144 ft. x 38 in) 70 gal./Bale 1 Ea.
Each for $341.00
Add to Cart
17-111-189
View Documents
Brady™
SPC152
43.8 m x 48.2 cm (144 ft. x 19 in.) 72 gal./Bale 2/Cs.
Case of 2 for $315.00
Add to Cart
Description
Description
  • Single-ply construction with MAXX technology
  • Hydrophobic nature for outdoor applications
  • Floats indefinitely even when saturated with oil
  • Dimpled and perforated
  • Heavier and bonded to reduce linting
  • Available in various sizes
Specifications
Specifications
MAXX Enhanced Heavy Weight Roll
Overspray leaks and drips, workstations, drums - liquid storage, outfalls and discharge Ponds