Learn More
DWK Life Sciences Wheaton™ Glass Diagnostic Vials with Screw Threads
Alternative to serum bottles and vials
$1070.00 - $1369.00
Catalog Number | Mfr. No. | Volume (Metric) | Volume (English) | Color | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Volume (English) | Color | Price | Quantity & Availability | |||
02-912-452
|
DWK Life Sciences
W219366 |
10 mL | 0.34 | Clear |
Case of 480 for $1,223.00
|
|
|||
02-912-451
|
DWK Life Sciences
W216375 |
5 mL | 0.17 | Amber |
Case of 480 for $1,258.00
|
|
|||
02-912-453
|
DWK Life Sciences
W216376 |
10 mL | 0.34 | Amber |
Case of 480 for $1,217.00
|
|
|||
02-912-457
|
DWK Life Sciences
W216378 |
30 mL | 1.01 | Amber |
Case of 288 for $1,070.00
|
|
|||
02-912-455
|
DWK Life Sciences
W216377 |
20 mL | 0.68 | Amber |
Case of 480 for $1,369.00
|
|
|||
02-912-450
|
DWK Life Sciences
W219365 |
5 mL | 0.17 | Clear |
Case of 480 for $1,116.00
|
|
|||
Description
Ideal for lyophilization.
- Screw-thread design eliminates the need for crimping
- Fits with I-Loc™ closure or screw cap with thin flange stopper
- Tubular design provides excellent clarity and dimensional consistency from vial to vial
- Heavy-walled glass provides impact resistance
Specifications
Type 1 | |
Screw | |
Diagnostic Vial |