Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Excelta™ Serrated Locking Hemostat w/Plastic Handle

Has vinyl-coated handles

Manufacturer:  Excelta™ 39PH

 View more versions of this product

Catalog No. 18-100-966


Add to cart

Description

Description

  • Available in Straight/Curved shape
  • Made of stainless steel
  • Overall length: 200mm (8 in.)

Specifications

Specifications

200mm
Straight
Locking Hemostat
8 in.
Stainless steel
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit