Learn More
SeraCare™ AccuSpan™ Linearity Panels
For evaluating and monitoring molecular diagnostic assay procedures used for detection of viral nucleic acids
Manufacturer:  SeraCare™ 24100254
 View more versions of this product
Catalog No. 22-554-023
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
  • Each panel consists of four members: one negative, one low positive, one medium positive and one high positive sample
  • Comes in ready-to-go, single-use format with target concentrations that cover range of most molecular assays
  • Full size: 4 x 1mL
Specifications
Specifications
BK Virus (BKV)
1 negative, 1 low positive, 1 medium positive, 1 high positive sample
4 x 1mL