Learn More
Cole-Parmer Monarch DC2C1-U4-11000 4-Channel Paperless Recorder; Color/6 Relay  DFS Item
Manufacturer:  Cole-Parmer 4900578
FEATURES
  • 2, 4, 6 or 12 direct universal inputs: DC voltage, current, thermocouples, RTD and dry contact
  • Serial output RS232 or RS485 (optional)
  • Ethernet 10 Base T option available
  • High speed storage capability: up to 8 samples per second/channel
  • Storage Media Compact Flash Card
  • 5.6" Brilliant TFT active matrix color display or 5" monochrome display
  • Intuitive Touchscreen Control with anti-glare coating
  • 6 relay contact or solid state relay outputs/3 digital control input
  • Parallel printer output (optional)
Catalog No. 50-631-732