Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4R,5R)-1,3-Dimethyl-4,5-diphenyl-2-[(S)-1-benzyl-2-hydroxyethylimino]imidazolidine 98.0+%, TCI America™

Click to view available options
Quantity:
100mg
337308-63-1
C26H29N3O
399.538
MFCD06797085
RAFFEHDNCQXIQG-ISJGIBHGSA-N
11177037
(2S)-2-[[(4R,5R)-1,3-dimethyl-4,5-diphenylimidazolidin-2-ylidene]amino]-3-phenylpropan-1-ol
CN1C(C(N(C1=NC(CC2=CC=CC=C2)CO)C)C3=CC=CC=C3)C4=CC=CC=C4
337308-63-1
C26H29N3O
399.538
MFCD06797085
RAFFEHDNCQXIQG-ISJGIBHGSA-N
11177037
(2S)-2-[[(4R,5R)-1,3-dimethyl-4,5-diphenylimidazolidin-2-ylidene]amino]-3-phenylpropan-1-ol
CN1C(C(N(C1=NC(CC2=CC=CC=C2)CO)C)C3=CC=CC=C3)C4=CC=CC=C4
Melting Point 114°C
Color White
Quantity 100mg
Formula Weight 399.54
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC,N)
Chemical Name or Material (4R,5R)-1,3-Dimethyl-4,5-diphenyl-2-[(S)-1-benzyl-2-hydroxyethylimino]imidazolidine