Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(4R,5S)-(+)-4-Methyl-5-phenyl-2-oxazolidinone, 99%, ACROS Organics™
Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
Specifications
Melting Point | 118°C to 121°C |
Specific Rotation | + 162 |
Specific Rotation Condition | + 162.00 (20.00°C c=2,CHCL3) |
Infrared Spectrum | Authentic |
Color | Brown-Yellow to Colorless |
Packaging | Glass bottle |
Quantity | 5g |
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Assay Percent Range | 98.5% min. (HPLC) |
Show More |