Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4R,6R)-tert-Butyl-6-(2-aminoethyl)-2,2-dimethyl-1,3-dioxane-4-acetate, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
125995-13-3
C14H27NO4
273.37
MFCD07369252
HWSHVKNLMBMKSR-GHMZBOCLSA-N
4r,6r-tert-butyl-6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate,tert-butyl 2-4r,6r-6-2-aminoethyl-2,2-dimethyl-1,3-dioxan-4-yl acetate,4r,6r-t-butyl-6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate,4r,cis-1,1-dimethylethyl-6-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate,4r,6r-t-butyl-6-2-aminoethyl-2,2-dimethyl-1,3,4r,6r-tert-butyl-6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4acetate,+/--cis-1,1-dimethylethyl 6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate,4r-2,2-dimethyl-6beta-2-aminoethyl-1,3-dioxane-4beta-acetic acid tert-butyl ester,4r-cis-1,1-dimethyethyl 6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate
2734288
tert-butyl 2-[(4R,6R)-6-(2-aminoethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate
CC1(OC(CC(O1)CC(=O)OC(C)(C)C)CCN)C
125995-13-3
C14H27NO4
273.37
MFCD07369252
HWSHVKNLMBMKSR-GHMZBOCLSA-N
4r,6r-tert-butyl-6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate,tert-butyl 2-4r,6r-6-2-aminoethyl-2,2-dimethyl-1,3-dioxan-4-yl acetate,4r,6r-t-butyl-6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate,4r,cis-1,1-dimethylethyl-6-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate,4r,6r-t-butyl-6-2-aminoethyl-2,2-dimethyl-1,3,4r,6r-tert-butyl-6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4acetate,+/--cis-1,1-dimethylethyl 6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate,4r-2,2-dimethyl-6beta-2-aminoethyl-1,3-dioxane-4beta-acetic acid tert-butyl ester,4r-cis-1,1-dimethyethyl 6-2-aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate
2734288
tert-butyl 2-[(4R,6R)-6-(2-aminoethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate
CC1(OC(CC(O1)CC(=O)OC(C)(C)C)CCN)C
Packaging Glass bottle
Quantity 1g
Color Blue or Green to Yellow
Refractive Index 1.4540 to 1.4560
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 273.37
Physical Form Viscous Liquid
Percent Purity 95%
Chemical Name or Material (4R,6R)-tert-Butyl-6-(2-aminoethyl)-2,2-dimethyl-1,3-dioxane-4-acetate, 95%
Show More Show Less