Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4R)-N-(tert-Butyldimethylsilyl)azetidin-2-one-4-carboxylic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
162856-35-1
C10H19NO3Si
229.35
LIEWITJXZYCDLE-SSDOTTSWSA-N
r-1-tert-butyldimethylsilyl-4-oxoazetidine-2-carboxylic acid,2r-1-tert-butyldimethylsilyl-4-oxoazetidine-2-carboxylic acid,4r-n-tert-butyldimethylsilyl azetidin-2-one-carboxylic acid,4r-n-tert-butyldimethylsilyl azetidin-2-one-4-carboxylic acid,1-tert-butyldimethylsilyl-4-oxoazetidine-2 r-carboxylic acid,2r-1-tert-butyl dimethyl silyl-4-oxoazetidine-2-carboxylic acid,r-1-tert-butyl-dimethyl-silanyl-4-oxo-azetidine-2-carboxylic acid,2-azetidinecarboxylicacid, 1-1,1-dimethylethyl dimethylsilyl-4-oxo-, 2r,2-azetidinecarboxylicacid,1-1,1-dimethylethyl dimethylsilyl-4-oxo-, 2r
2733822
(2R)-1-[tert-butyl(dimethyl)silyl]-4-oxoazetidine-2-carboxylic acid
CC(C)(C)[Si](C)(C)N1C(CC1=O)C(=O)O
162856-35-1
C10H19NO3Si
229.35
LIEWITJXZYCDLE-SSDOTTSWSA-N
r-1-tert-butyldimethylsilyl-4-oxoazetidine-2-carboxylic acid,2r-1-tert-butyldimethylsilyl-4-oxoazetidine-2-carboxylic acid,4r-n-tert-butyldimethylsilyl azetidin-2-one-carboxylic acid,4r-n-tert-butyldimethylsilyl azetidin-2-one-4-carboxylic acid,1-tert-butyldimethylsilyl-4-oxoazetidine-2 r-carboxylic acid,2r-1-tert-butyl dimethyl silyl-4-oxoazetidine-2-carboxylic acid,r-1-tert-butyl-dimethyl-silanyl-4-oxo-azetidine-2-carboxylic acid,2-azetidinecarboxylicacid, 1-1,1-dimethylethyl dimethylsilyl-4-oxo-, 2r,2-azetidinecarboxylicacid,1-1,1-dimethylethyl dimethylsilyl-4-oxo-, 2r
2733822
(2R)-1-[tert-butyl(dimethyl)silyl]-4-oxoazetidine-2-carboxylic acid
CC(C)(C)[Si](C)(C)N1C(CC1=O)C(=O)O
Packaging Glass Bottle
Melting Point 137.0°C to 141.0°C
Color White
Quantity 5g
Infrared Spectrum Authentic
Assay Percent Range 99%
Formula Weight 229.35
Physical Form Crystalline Powder or Chunks
Percent Purity 99%
Chemical Name or Material (4R)-N-(tert-Butyldimethylsilyl)azetidin-2-one-4-carboxylic acid