Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4S)-(-)-4,5-Dihydro-2-[2'-(diphenylphosphino)phenyl]-4-isopropyloxazole, 98%, ACROS Organics™

Click to view available options
Quantity:
100mg
500mg
Packaging:
Glass bottle
148461-14-7
C24H24NOP
373.436
MFCD03427599
OUQSAXROROGQEE-JOCHJYFZSA-N
s-ipr-phox,s---2-2-diphenylphosphino phenyl-4-isopropyl-2-oxazoline,s-2-2-diphenylphosphino phenyl-4-isopropyl-4,5-dihydrooxazole,2-2-diphenylphosphino phenyl-4beta-isopropyl-2-oxazoline,-4s-2-2-diphenylphosphino phenyl-4-2-propyl oxazolin,--4s-2-2-diphenylphosphino phenyl-4-2-propyl oxazoline,s---2-2-diphenylphosphino phenyl-4,5-dihydro-4-isopropyloxazole,s---2-2-diphenylphosphino phenyl-4-1-isopropyl-4,5-dihydrooxazole,s---2-2-diphenylphosphino phenyl-4-isopropyl-2-oxazoline chn
9807779
diphenyl-[2-[(4S)-4-propan-2-yl-4,5-dihydro-1,3-oxazol-2-yl]phenyl]phosphane
CC(C)C1COC(=N1)C2=CC=CC=C2P(C3=CC=CC=C3)C4=CC=CC=C4
148461-14-7
C24H24NOP
373.436
MFCD03427599
OUQSAXROROGQEE-JOCHJYFZSA-N
s-ipr-phox,s---2-2-diphenylphosphino phenyl-4-isopropyl-2-oxazoline,s-2-2-diphenylphosphino phenyl-4-isopropyl-4,5-dihydrooxazole,2-2-diphenylphosphino phenyl-4beta-isopropyl-2-oxazoline,-4s-2-2-diphenylphosphino phenyl-4-2-propyl oxazolin,--4s-2-2-diphenylphosphino phenyl-4-2-propyl oxazoline,s---2-2-diphenylphosphino phenyl-4,5-dihydro-4-isopropyloxazole,s---2-2-diphenylphosphino phenyl-4-1-isopropyl-4,5-dihydrooxazole,s---2-2-diphenylphosphino phenyl-4-isopropyl-2-oxazoline chn
9807779
diphenyl-[2-[(4S)-4-propan-2-yl-4,5-dihydro-1,3-oxazol-2-yl]phenyl]phosphane
CC(C)C1COC(=N1)C2=CC=CC=C2P(C3=CC=CC=C3)C4=CC=CC=C4
Quantity 500mg
Packaging Glass bottle
Color White
Specific Rotation -43° to -46° (20°C, 589nm) (C=1, CHCl3)
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 373.43
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material (4S)-(-)-4, 5-Dihydro-2-[2'-(diphenylphosphino)phenyl]-4-isopropyloxazole
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.