Alfa Aesar™ (4S,5S)-(+)-4,5-Bis(fluoromethyl)-2,2-dimethyl-1,3-dioxolane, 90+%

Manufacturer: Alfa Aesar™ H2776603

Quantity 1g

 View more versions of this product

Catalog No. AAH2776603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 84409-70-1
    Molecular Formula C7H12F2O2
    Formula Weight 166.17
    MDL Number MFCD01074597
    Synonym (+)-2,3-O-Isopropylidiene-1,4-difluoro-2,3-butanediol
    Chemical Name or Material (4S,5S)-(+)-4,5-Bis(fluoromethyl)-2,2-dimethyl-1,3-dioxolane

    Specifications

    Formula Weight 166.17
    Synonym (+)-2,3-O-Isopropylidiene-1,4-difluoro-2,3-butanediol
    Molecular Formula C7H12F2O2
    MDL Number MFCD01074597
    CAS 84409-70-1
    Quantity 1g
    Chemical Name or Material (4S,5S)-(+)-4,5-Bis(fluoromethyl)-2,2-dimethyl-1,3-dioxolane
    Assay ≥90%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit