Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4S)-N-Crotonyl-4-benzyl-2-oxazolidinone, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
90719-30-5
C14H15NO3
MFCD00278769
locostatin,s-4-benzyl-3-but-2-enoyl oxazolidin-2-one,s,e-4-benzyl-3-but-2-enoyl oxazolidin-2-one,4s-n-crotonyl-4-benzyl-2-oxazolidinone,4s-3-e-but-2-enoyl-4-benzyl-2-oxazolidinone,4s-4-benzyl-3-e-but-2-enoyl-1,3-oxazolidin-2-one,cell sheet migration inhibitor, locostatin,4s-4-benzyl-3-2e-but-2-enoyl-1,3-oxazolidin-2-one,locostatin hplc,3-crotonyl-4beta-benzyloxazolidine-2-one
90719-30-5
C14H15NO3
MFCD00278769
locostatin,s-4-benzyl-3-but-2-enoyl oxazolidin-2-one,s,e-4-benzyl-3-but-2-enoyl oxazolidin-2-one,4s-n-crotonyl-4-benzyl-2-oxazolidinone,4s-3-e-but-2-enoyl-4-benzyl-2-oxazolidinone,4s-4-benzyl-3-e-but-2-enoyl-1,3-oxazolidin-2-one,cell sheet migration inhibitor, locostatin,4s-4-benzyl-3-2e-but-2-enoyl-1,3-oxazolidin-2-one,locostatin hplc,3-crotonyl-4beta-benzyloxazolidine-2-one
Quantity 1g
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 245.28
Chemical Name or Material (4S)-N-Crotonyl-4-benzyl-2-oxazolidinone, 99%