Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4S)-N-Crotonyl-4-isopropyl-2-oxazolidinone, 99%, Thermo Scientific™

Catalog Number AC337790010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
90719-29-2
C10H15NO3
197.23
MFCD00075429
YZRHIMNEVFYTAJ-WTSVBCDHSA-N
4s-3-2e-but-2-enoyl-4-isopropyl-1,3-oxazolidin-2-one, hg^l@lanry gyifjffjamdp, 4s-n-crotonyl-4-isopropyl-2-oxazolidinone, 3-2-butenoyl-4beta-isopropyloxazolidine-2-one, 4s-3-crotonoyl-4beta-isopropyloxazolidine-2-one, s-+-3-crotonoyl-4-isopropyl-2-oxazolidinone, 4s-3-e-but-2-enoyl-4-propan-2-yl-1,3-oxazolidin-2-one, 3-trans-3-methyl-2-propenoyl-4-s-isopropyl-2-oxazolidinone
5702601
(4S)-3-[(E)-but-2-enoyl]-4-propan-2-yl-1,3-oxazolidin-2-one
CC=CC(=O)N1C(COC1=O)C(C)C
90719-29-2
C10H15NO3
197.23
MFCD00075429
YZRHIMNEVFYTAJ-WTSVBCDHSA-N
4s-3-2e-but-2-enoyl-4-isopropyl-1,3-oxazolidin-2-one, hg^l@lanry gyifjffjamdp, 4s-n-crotonyl-4-isopropyl-2-oxazolidinone, 3-2-butenoyl-4beta-isopropyloxazolidine-2-one, 4s-3-crotonoyl-4beta-isopropyloxazolidine-2-one, s-+-3-crotonoyl-4-isopropyl-2-oxazolidinone, 4s-3-e-but-2-enoyl-4-propan-2-yl-1,3-oxazolidin-2-one, 3-trans-3-methyl-2-propenoyl-4-s-isopropyl-2-oxazolidinone
5702601
(4S)-3-[(E)-but-2-enoyl]-4-propan-2-yl-1,3-oxazolidin-2-one
CC=CC(=O)N1C(COC1=O)C(C)C
CAS Min % 98.5
CAS Max % 100.0
Melting Point 55°C to 58°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation + 102.5
Formula Weight 197.23
Percent Purity 99%
Physical Form Crystals
Chemical Name or Material (4S)-N-Crotonyl-4-isopropyl-2-oxazolidinone, 99%
Show More Show Less

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit