Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4S)-N-(tert-Butyldimethylsilyl)azetidin-2-one-4-carboxylic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
250mg
Packaging:
Glass Bottle
82938-50-9
C10H19NO3Si
229.351
MFCD01862126
LIEWITJXZYCDLE-ZETCQYMHSA-N
4s-n-tert-butyldimethylsilyl azetidin-2-one-4-carboxylic acid,s-1-tert-butyldimethylsilyl-4-oxoazetidine-2-carboxylic acid,2s-1-tert-butyl dimethyl silyl-4-oxo-azetidine-2-carboxylic acid,2s-1-tert-butyldimethylsilyl-4-oxoazetidine-2-carboxylic acid,2s-1-tert-butyl dimethyl silyl-4-oxoazetidine-2-carboxylic acid,4s-n-tert-butyldimethylsilyl azetidin-2-one-carboxylic acid,4s-1-t-butyldimethylsilylazetidin-2-one-4-carboxylic acid,s-1-t-butyldimethylsilyl-4-oxo-2-azetidinecarboxylic acid,s-n-tert-butyldimethylsilyl-4-oxo-azetidin-2 carboxylic acid,2s-4-oxo-1-1,1,2,2-tetramethyl-1-silapropyl azetidine-2-carboxylic acid
10489602
(2S)-1-[tert-butyl(dimethyl)silyl]-4-oxoazetidine-2-carboxylic acid
CC(C)(C)[Si](C)(C)N1C(CC1=O)C(=O)O
82938-50-9
C10H19NO3Si
229.351
MFCD01862126
LIEWITJXZYCDLE-ZETCQYMHSA-N
4s-n-tert-butyldimethylsilyl azetidin-2-one-4-carboxylic acid,s-1-tert-butyldimethylsilyl-4-oxoazetidine-2-carboxylic acid,2s-1-tert-butyl dimethyl silyl-4-oxo-azetidine-2-carboxylic acid,2s-1-tert-butyldimethylsilyl-4-oxoazetidine-2-carboxylic acid,2s-1-tert-butyl dimethyl silyl-4-oxoazetidine-2-carboxylic acid,4s-n-tert-butyldimethylsilyl azetidin-2-one-carboxylic acid,4s-1-t-butyldimethylsilylazetidin-2-one-4-carboxylic acid,s-1-t-butyldimethylsilyl-4-oxo-2-azetidinecarboxylic acid,s-n-tert-butyldimethylsilyl-4-oxo-azetidin-2 carboxylic acid,2s-4-oxo-1-1,1,2,2-tetramethyl-1-silapropyl azetidine-2-carboxylic acid
10489602
(2S)-1-[tert-butyl(dimethyl)silyl]-4-oxoazetidine-2-carboxylic acid
CC(C)(C)[Si](C)(C)N1C(CC1=O)C(=O)O
Quantity 250mg
Packaging Glass Bottle
Melting Point 135.0°C to 137.0°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 229.35
Percent Purity 99%
Chemical Name or Material (4S)-N-(tert-Butyldimethylsilyl)azetidin-2-one-4-carboxylic acid, 99%