Learn More
Medchemexpress LLC 4SC-203 | 895533-09-2 | MFCD28502039 | 99.9% | 642.77 | C33H38N8O4S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1989710MG
Encompass_Preferred
4SC-203 is a small-molecule multikinase inhibitor investigated for antineoplastic activity. It selectively targets FMS-related tyrosine kinase 3 (FLT3), including mutant forms, and vascular endothelial growth factor receptors (VEGFRs), and has been used in preclinical and early clinical research settings. Supplied as a high-purity research reagent for laboratory studies.
  • Selective inhibition of FLT3, including mutant forms.
  • Inhibits vascular endothelial growth factor receptors.
  • Demonstrated antineoplastic activity in preclinical studies.
  • Suitable for in vitro and in vivo research use (not for human use).
  • High purity appropriate for research applications.
  • Well characterized with CAS and chemical identifiers for traceability.
Catalog No. 50-002-97053