Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,11,17,23-Tetrakis-chloromethyl-25,26,27,28-tetrapropoxy-calix[4]arene, 98%, ACROS Organics™

Click to view available options
Quantity:
500mg
Packaging:
Glass Bottle
325814-49-1
C44H52Cl4O4
786.696
MFCD02100943
XCYJNZCPDHLBRV-UHFFFAOYSA-N
5,11,17,23-chloromethyl-25,26,27,28-tetrapropoxy-calix 4 arene
2733873
CCCOC1=C2CC3=CC(=CC(=C3OCCC)CC4=C(C(=CC(=C4)CCl)CC5=C(C(=CC(=C5)CCl)CC1=CC(=C2)CCl)OCCC)OCCC)CCl
325814-49-1
C44H52Cl4O4
786.696
MFCD02100943
XCYJNZCPDHLBRV-UHFFFAOYSA-N
5,11,17,23-chloromethyl-25,26,27,28-tetrapropoxy-calix 4 arene
2733873
CCCOC1=C2CC3=CC(=CC(=C3OCCC)CC4=C(C(=CC(=C4)CCl)CC5=C(C(=CC(=C5)CCl)CC1=CC(=C2)CCl)OCCC)OCCC)CCl
Chelation/Complexation reagent
Packaging Glass Bottle
Infrared Spectrum Authentic
Quantity 500mg
Melting Point 218.0°C to 224.0°C
Color White to Cream
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 786.71
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 5,11,17,23-Tetrakis-chloromethyl-25,26,27,28-tetrapropoxy-calix[4]arene
Show More Show Less