Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,11,17,23-Tetrakis-mercaptomethyl-25,26,27,28-tetrapropoxycalix[4]arene, 98%, Acros Organics

Click to view available options
Quantity:
1g
325776-17-8
C44H56O4S4
777.168
MFCD02100956
LDYAWJKFSRUXSQ-UHFFFAOYSA-N
5,11,17,23-tetrakis-mercaptomethyl-25,26,27,28-tetrapropoxycalix 4!arene
118856128
CCCOC1=C2C=CC=C1C(C3=C(C(=CC=C3)C(C4=CC=CC(=C4OCCC)C(C5=CC=CC(=C5OCCC)C2CS)CS)CS)OCCC)CS
325776-17-8
C44H56O4S4
777.168
MFCD02100956
LDYAWJKFSRUXSQ-UHFFFAOYSA-N
5,11,17,23-tetrakis-mercaptomethyl-25,26,27,28-tetrapropoxycalix 4!arene
118856128
CCCOC1=C2C=CC=C1C(C3=C(C(=CC=C3)C(C4=CC=CC(=C4OCCC)C(C5=CC=CC(=C5OCCC)C2CS)CS)CS)OCCC)CS
Chelation/Complexation reagent
Quantity 1g
Color White
Melting Point 194°C to 200°C
Infrared Spectrum Authentic
Elemental Analysis 97.5% min.
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 777.2
Physical Form Crystalline Powder and Chunks
Chemical Name or Material 5,11,17,23-Tetrakis-mercaptomethyl-25,26,27,28-tetrapropoxycalix[4]arene
Show More Show Less