Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-(2-Bromophenyl)-1H-tetrazole, 98+%

Click to view available options
Quantity:
1g
5g
25g
73096-42-1
C7H5BrN4
225.049
MFCD00151799
YHVBXKTXLJTDRI-UHFFFAOYSA-N
5-2-bromophenyl-1h-tetrazole,5-2-bromophenyl tetrazole,5-2-bromo-phenyl-2h-tetrazole,5-2-bromophenyl-2h-tetrazole,5-2-bromophenyl-1h-1,2,3,4-tetrazole,5-2-bromophenyl-2h-1,2,3,4-tetrazole,5-2-bromophenyl-1h-tetraazole,5-2-bromophenyl-1 h-tetrazole,1h-tetrazole, 5-2-bromophenyl
560689
5-(2-bromophenyl)-2H-tetrazole
C1=CC=C(C(=C1)C2=NNN=N2)Br
73096-42-1
C7H5BrN4
225.049
MFCD00151799
YHVBXKTXLJTDRI-UHFFFAOYSA-N
5-2-bromophenyl-1h-tetrazole,5-2-bromophenyl tetrazole,5-2-bromo-phenyl-2h-tetrazole,5-2-bromophenyl-2h-tetrazole,5-2-bromophenyl-1h-1,2,3,4-tetrazole,5-2-bromophenyl-2h-1,2,3,4-tetrazole,5-2-bromophenyl-1h-tetraazole,5-2-bromophenyl-1 h-tetrazole,1h-tetrazole, 5-2-bromophenyl
560689
5-(2-bromophenyl)-2H-tetrazole
C1=CC=C(C(=C1)C2=NNN=N2)Br
Melting Point 181°C to 183°C
Quantity 1g
Beilstein 148567
Formula Weight 225.05
Percent Purity ≥98%
Chemical Name or Material 5-(2-Bromophenyl)-1H-tetrazole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning