Alfa Aesar™ 5-(2-Bromophenyl)isoxazole, 95%

Manufacturer: Alfa Aesar™ H3417109

Quantity 10g

 View more versions of this product

Catalog No. AAH3417109

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 387358-52-3
    Molecular Formula C9H6BrNO
    Formula Weight 224.06
    MDL Number MFCD02183530
    Chemical Name or Material 5-(2-Bromophenyl)isoxazole

    Specifications

    Formula Weight 224.06
    Molecular Formula C9H6BrNO
    MDL Number MFCD02183530
    CAS 387358-52-3
    Boiling Point 95° to 97°C (0.23mmHg)
    Quantity 10g
    Chemical Name or Material 5-(2-Bromophenyl)isoxazole
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit