Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(2-nitrophenyl)furfural, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
20000-96-8
C11H7NO4
217.18
MFCD00216586
QBYRUURYXPVDAK-UHFFFAOYSA-N
5-2-nitrophenyl-2-furaldehyde,5-2-nitrophenyl furan-2-carbaldehyde,5-2-nitrophenyl furfural,2-furancarboxaldehyde, 5-2-nitrophenyl,2-furancarboxaldehyde,5-2-nitrophenyl,5-2-nitro-phenyl-furan-2-carbaldehyde,acmc-1cjo1,cambridge id 7184607,5 2-nitro phenyl-furan-2-carbaldehyde
673649
5-(2-nitrophenyl)furan-2-carbaldehyde
C1=CC=C(C(=C1)C2=CC=C(O2)C=O)[N+](=O)[O-]
20000-96-8
C11H7NO4
217.18
MFCD00216586
QBYRUURYXPVDAK-UHFFFAOYSA-N
5-2-nitrophenyl-2-furaldehyde,5-2-nitrophenyl furan-2-carbaldehyde,5-2-nitrophenyl furfural,2-furancarboxaldehyde, 5-2-nitrophenyl,2-furancarboxaldehyde,5-2-nitrophenyl,5-2-nitro-phenyl-furan-2-carbaldehyde,acmc-1cjo1,cambridge id 7184607,5 2-nitro phenyl-furan-2-carbaldehyde
673649
5-(2-nitrophenyl)furan-2-carbaldehyde
C1=CC=C(C(=C1)C2=CC=C(O2)C=O)[N+](=O)[O-]
Color Orange to Brown
Melting Point 92°C to 94°C
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 5g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 217.18
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 5-(2-Nitrophenyl)furfural
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.