Learn More
Alfa Aesar™ 5-(3-Methoxyphenyl)isoxazole, 95%
Manufacturer:  Alfa Aesar™ H3396503
Quantity 1g
 View more versions of this product
Catalog No. AAH3396503
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
C10H9NO2
175.19
MFCD13176591
5-(3-Methoxyphenyl)isoxazole
Specifications
C10H9NO2
MFCD13176591
1g
95%
175.19
115° to 117°C (0.46mmHg)
5-(3-Methoxyphenyl)isoxazole