Learn More
Alfa Aesar™ 5-(3-Nitrophenyl)-2-furoyl chloride
Manufacturer:  Alfa Aesar™ H5040706
Quantity 5g
 View more versions of this product
Catalog No. AAH5040706
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Chemical Identifiers
61941-87-5
251.63
UN3261
C11H6ClNO4
MFCD02258037
5-(3-Nitrophenyl)-2-furoyl chloride
Specifications
5g
C11H6ClNO4
MFCD02258037
5-(3-Nitrophenyl)-2-furoyl chloride
61941-87-5
251.63
UN3261
Safety and Handling
SDS & Safety Documents
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.