Alfa Aesar™ 5-(4-Bromophenyl)isoxazole, 98%

Manufacturer: Alfa Aesar™ H5034203

Quantity 1g

 View more versions of this product

Catalog No. AAH5034203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7064-31-5
    Molecular Formula C9H6BrNO
    Formula Weight 224.06
    MDL Number MFCD02183532
    Chemical Name or Material 5-(4-Bromophenyl)isoxazole

    Specifications

    Formula Weight 224.06
    Melting Point 115° to 118°C
    Quantity 1g
    Molecular Formula C9H6BrNO
    MDL Number MFCD02183532
    CAS 7064-31-5
    Chemical Name or Material 5-(4-Bromophenyl)isoxazole
    Assay 98%

    Description

    Applications
    5-(4-Bromophenyl)isoxazole is used as an active pharmaceutical ingredients.

    Notes
    Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit