Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(4-Bromophenyl)isoxazole, 98%, Thermo Scientific Chemicals

Catalog Number AAH5034206
Click to view available options
Quantity:
5 g
7064-31-5
C9H6BrNO
224.057
MFCD02183532
AVMMSHZGFRWLRI-UHFFFAOYSA-N
5-4-bromophenyl isoxazole, 5-4-bromophenyl-1,2-oxazole, acmc-209ofh
2735608
5-(4-bromophenyl)-1,2-oxazole
C1=CC(=CC=C1C2=CC=NO2)Br
7064-31-5
C9H6BrNO
224.057
MFCD02183532
AVMMSHZGFRWLRI-UHFFFAOYSA-N
5-4-bromophenyl isoxazole, 5-4-bromophenyl-1,2-oxazole, acmc-209ofh
2735608
5-(4-bromophenyl)-1,2-oxazole
C1=CC(=CC=C1C2=CC=NO2)Br

5-(4-Bromophenyl)isoxazole is used as an active pharmaceutical ingredients.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-(4-Bromophenyl)isoxazole is used as an active pharmaceutical ingredients.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Melting Point 115°C to 118°C
Quantity 5 g
Formula Weight 224.06
Percent Purity 98%
Chemical Name or Material 5-(4-Bromophenyl)isoxazole
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit