Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 5-(4-Hydroxyphenyl)pentanoic acid, 98%
Click to view available options
Quantity:
1g
5g
25g
Description
Applications
5-(4-Hydroxyphenyl)pentanoic acid is used as pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
5-(4-Hydroxyphenyl)pentanoic acid is used as pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Specifications
Melting Point | 116°C to 120°C |
Quantity | 1g |
Solubility Information | Soluble in water. |
Formula Weight | 194.23 |
Percent Purity | 98% |
Chemical Name or Material | 5-(4-Hydroxyphenyl)pentanoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures