Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(4-Methylphenyl)-5-phenylhydantoin, 99+%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
51169-17-6
C16H14N2O2
266.29
MFCD00005263
WPAPSGQWYNPWCZ-UHFFFAOYSA-N
5-p-methylphenyl-5-phenylhydantoin,5-phenyl-5-p-tolyl hydantoin,5-phenyl-5-4-tolyl hydantoin,5-4-methylphenyl-5-phenylimidazolidine-2,4-dione,5-4-methylphenyl-5-phenyl-2,4-imidazolidinedione,5-p-tolyl-5-phenylhydantoin,5-phenyl-5-p-tolyl imidazolidine-2,4-dione,2,4-imidazolidinedione, 5-4-methylphenyl-5-phenyl,5-4-methylphenyl-5-phenylhydantoin,5-4-methylphenyl-5-phenyl hydantoin
92334
5-(4-methylphenyl)-5-phenylimidazolidine-2,4-dione
CC1=CC=C(C=C1)C2(C(=O)NC(=O)N2)C3=CC=CC=C3
51169-17-6
C16H14N2O2
266.29
MFCD00005263
WPAPSGQWYNPWCZ-UHFFFAOYSA-N
5-p-methylphenyl-5-phenylhydantoin,5-phenyl-5-p-tolyl hydantoin,5-phenyl-5-4-tolyl hydantoin,5-4-methylphenyl-5-phenylimidazolidine-2,4-dione,5-4-methylphenyl-5-phenyl-2,4-imidazolidinedione,5-p-tolyl-5-phenylhydantoin,5-phenyl-5-p-tolyl imidazolidine-2,4-dione,2,4-imidazolidinedione, 5-4-methylphenyl-5-phenyl,5-4-methylphenyl-5-phenylhydantoin,5-4-methylphenyl-5-phenyl hydantoin
92334
5-(4-methylphenyl)-5-phenylimidazolidine-2,4-dione
CC1=CC=C(C=C1)C2(C(=O)NC(=O)N2)C3=CC=CC=C3
Melting Point 225.0°C to 228.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Color White
Quantity 1g
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Formula Weight 266.29
Physical Form Powder
Percent Purity 99+%
Chemical Name or Material 5-(4-Methylphenyl)-5-phenylhydantoin, 99+%
Show More Show Less

EINECSNumber : 257-028-3