Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(4-Morpholinyl)-2-nitrophenol, 97%, Thermo Scientific Chemicals

Catalog Number AAB2495003
Click to view available options
Quantity:
1 g
5 g
175135-19-0
C10H12N2O4
224.216
MFCD00053057
XXIIDARBNMZFEH-UHFFFAOYSA-N
5-morpholino-2-nitrophenol, 5-4-morpholinyl-2-nitrophenol, 5-4-morpholino-2-nitrophenol, 5-morpholin-4-yl-2-nitrophenol, 5-morpholin-4-yl-2-nitro-phenolate, maybridge1_001296, acmc-1c6gm, 2-nitro-5-morpholinophenol, 5-morpholin-4-yl-2-nitro-phenol, n-3-hydroxy-4-nitrophenyl morpholine
2799399
5-morpholin-4-yl-2-nitrophenol
C1COCCN1C2=CC(=C(C=C2)[N+](=O)[O-])O
175135-19-0
C10H12N2O4
224.216
MFCD00053057
XXIIDARBNMZFEH-UHFFFAOYSA-N
5-morpholino-2-nitrophenol, 5-4-morpholinyl-2-nitrophenol, 5-4-morpholino-2-nitrophenol, 5-morpholin-4-yl-2-nitrophenol, 5-morpholin-4-yl-2-nitro-phenolate, maybridge1_001296, acmc-1c6gm, 2-nitro-5-morpholinophenol, 5-morpholin-4-yl-2-nitro-phenol, n-3-hydroxy-4-nitrophenyl morpholine
2799399
5-morpholin-4-yl-2-nitrophenol
C1COCCN1C2=CC(=C(C=C2)[N+](=O)[O-])O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 143°C to 145°C
Quantity 1 g
Formula Weight 224.22
Percent Purity 97%
Chemical Name or Material 5-(4-Morpholinyl)-2-nitrophenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit