Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(4-Nitrophenyl)-2-furoic acid, ACROS Organics™

Click to view available options
Quantity:
1g
5g
28123-73-1
C11H7NO5
233.18
MFCD00094261
FXZTYDPWMQBTDH-UHFFFAOYSA-N
5-4-nitrophenyl furan-2-carboxylic acid,5-4-nitrophenyl-2-furoic acid,5-4-nitro-phenyl-furan-2-carboxylic acid,2-furancarboxylic acid, 5-4-nitrophenyl,2-furancarboxylic acid,5-4-nitrophenyl,5-p-nitrophenyl-2-furancarboxylic acid,5-4-nitrophenyl-2-furancarboxylic acid,5-4-nitrophenyl-2-furan carboxylic acid
161593
C1=CC(=CC=C1C2=CC=C(O2)C(=O)O)[N+](=O)[O-]
28123-73-1
C11H7NO5
233.18
MFCD00094261
FXZTYDPWMQBTDH-UHFFFAOYSA-N
5-4-nitrophenyl furan-2-carboxylic acid,5-4-nitrophenyl-2-furoic acid,5-4-nitro-phenyl-furan-2-carboxylic acid,2-furancarboxylic acid, 5-4-nitrophenyl,2-furancarboxylic acid,5-4-nitrophenyl,5-p-nitrophenyl-2-furancarboxylic acid,5-4-nitrophenyl-2-furancarboxylic acid,5-4-nitrophenyl-2-furan carboxylic acid
161593
C1=CC(=CC=C1C2=CC=C(O2)C(=O)O)[N+](=O)[O-]
Packaging Glass Bottle
Melting Point 255°C to 257°C
Quantity 5g
Formula Weight 233.18
Chemical Name or Material 5-(4-Nitrophenyl)-2-furoic acid

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Risk of explosion if heated under confinement.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning